Tisk…

Methyl 2-chloro-6-morpholin-4-ylisonicotinate

Dodavatel: Apollo Scientific

OR110252-1G OR110252-250MG
APOSOR110252-1GEA 5860 CZK
APOSOR110252-1G APOSOR110252-250MG
Methyl 2-chloro-6-morpholin-4-ylisonicotinate
Methyl 2-chloro-6-morpholin-4-ylisonicotinate
Vzorec: C₁₁H₁₃ClN₂O₃
MW: 256,686 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12545988
CAS číslo: 1201675-09-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR