N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine
Dodavatel: Apollo Scientific
OR110239-1G
OR110239-250MG
APOSOR110239-1GEA
9920
CZK
APOSOR110239-1G
APOSOR110239-250MG
N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine
N,N-Diethyl-1,3,4-thiadiazole-2,5-diamine
Vzorec:
C₆H₁₂N₄S Storage Temperature: Ambient |
MDL Number:
MFCD12756249 CAS číslo: 894880-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...