Tisk…

2-Chloro-1-ethyl-1H-indole-3-carbaldehyde

Dodavatel: Apollo Scientific

OR110223-1G OR110223-250MG
APOSOR110223-1GEA 6620 CZK
APOSOR110223-1G APOSOR110223-250MG
2-Chloro-1-ethyl-1H-indole-3-carbaldehyde
2-Chloro-1-ethyl-1H-indole-3-carbaldehyde
Vzorec: C₁₁H₁₀ClNO
MW: 207,656 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02253188
CAS číslo: 64788-54-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR