5-Iodo-1-methyl-1H-indazol-3-amine
Dodavatel: Apollo Scientific
|
Varování
|
OR110216-1G
OR110216-5G
APOSOR110216-1GEA
6620
CZK
APOSOR110216-1G
APOSOR110216-5G
5-Iodo-1-methyl-1H-indazol-3-amine
5-Iodo-1-methyl-1H-indazol-3-amine
Vzorec:
C₈H₈IN₃ MW: 273.074 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11520131 CAS číslo: 1227955-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...