Ethyl 3-amino-5-iodo-1-benzothiophene-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR110214-1G
OR110214-5G
APOSOR110214-1GEA
6620
CZK
APOSOR110214-1G
APOSOR110214-5G
Ethyl 3-amino-5-iodo-1-benzothiophene-2-carboxylate
Ethyl 3-amino-5-iodo-1-benzothiophene-2-carboxylate
Vzorec:
C₁₁H₁₀INO₂S MW: 347,172 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21100196 CAS číslo: 1427461-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...