Ethyl 7-bromo-5-nitro-1-benzofuran-2-carboxylate
Dodavatel: Apollo Scientific
|
Varování
|
OR110176-1G
OR110176-5G
APOSOR110176-1GEA
2070
CZK
APOSOR110176-1G
APOSOR110176-5G
Ethyl 7-bromo-5-nitro-1-benzofuran-2-carboxylate
Ethyl 7-bromo-5-nitro-1-benzofuran-2-carboxylate
Vzorec:
C₁₁H₈BrNO₅ MW: 314,089 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11112161 CAS číslo: 1010072-35-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...