2-Methyl-6-nitro-2H-indazole
Dodavatel: Apollo Scientific
OR110174-1G
OR110174-5G
APOSOR110174-1GEA
4400
CZK
APOSOR110174-1G
APOSOR110174-5G
2-Methyl-6-nitro-2H-indazole
2-Methyl-6-nitro-2H-indazole
Vzorec:
C₈H₇N₃O₂ MW: 177,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00456075 CAS číslo: 6850-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...