Tisk…

4-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one

Dodavatel: Apollo Scientific

OR110159-1G OR110159-250MG
APOSOR110159-1GEA 5860 CZK
APOSOR110159-1G APOSOR110159-250MG
4-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
4-(3-Methoxyphenyl)-2,4-dihydro-3H-1,2,4-triazol-3-one
Vzorec: C₉H₉N₃O₂
MW: 191,187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22370030
CAS číslo: 1427460-99-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR