Tisk…

3-Chloro-5-methylimidazo[1,2-a]pyridine

Dodavatel: Apollo Scientific
Varování

OR110129-1G OR110129-250MG
APOSOR110129-1GEA 11410 CZK
APOSOR110129-1G APOSOR110129-250MG
3-Chloro-5-methylimidazo[1,2-a]pyridine
3-Chloro-5-methylimidazo[1,2-a]pyridine
Vzorec: C₈H₇ClN₂
MW: 166.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21336365
CAS číslo: 334711-68-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR