Tisk…

1-[1-(tert-Butoxycarbonyl)pyrrolidin-3-yl]piperidine-4-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR110106-1G OR110106-250MG
APOSOR110106-1GEA 10250 CZK
APOSOR110106-1G APOSOR110106-250MG
1-[1-(tert-Butoxycarbonyl)pyrrolidin-3-yl]piperidine-4-carboxylic acid
1-[1-(tert-Butoxycarbonyl)pyrrolidin-3-yl]piperidine-4-carboxylic acid
Vzorec: C₁₅H₂₆N₂O₄
MW: 298,378 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09909347
CAS číslo: 1440535-47-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR