2-Thioxo-2,3-dihydro-1,3-benzoxazole-5-carbonitrile
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR110092-100MG
OR110092-1G
APOSOR110092-100MGEA
8270
CZK
APOSOR110092-100MG
APOSOR110092-1G
2-Thioxo-2,3-dihydro-1,3-benzoxazole-5-carbonitrile
2-Thioxo-2,3-dihydro-1,3-benzoxazole-5-carbonitrile
Vzorec:
C₈H₄N₂OS MW: 176,195 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD24562615 CAS číslo: 93794-41-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...