Tisk…

(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid

Dodavatel: Apollo Scientific

OR110051-1G OR110051-5G
APOSOR110051-1GEA 5290 CZK
APOSOR110051-1G APOSOR110051-5G
(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
(2R,3S)-3-Hydroxy-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)butanoic acid
Vzorec: C₁₂H₁₃NO₄
MW: 235,236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24850048
CAS číslo: 1440531-71-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR