Tisk…

Dodecane-1,12-diamine, N-BOC protected 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (12-aminododecyl)carbamate, tert-Butyl (12-aminododec-1-yl)carbamate, 1,12-Diaminododecane, N-BOC protected, Boc-NH-C12-NH2

OR1098-100MG OR1098-250MG OR1098-1G
APOSOR1098-100MGEA 2070 CZK
APOSOR1098-100MG APOSOR1098-250MG APOSOR1098-1G
Dodecane-1,12-diamine, N-BOC protected 95%
Dodecane-1,12-diamine, N-BOC protected
Vzorec: C₁₇H₃₆N₂O₂
Bod tání: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094503
CAS číslo: 109792-60-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR