Tisk…

Decane-1,10-diamine, N-BOC protected 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl (10-aminodecyl)carbamate

OR1097-1G OR1097-250MG
APOSOR1097-1GEA 4570 CZK
APOSOR1097-1G APOSOR1097-250MG
Decane-1,10-diamine, N-BOC protected 97%
Decane-1,10-diamine, N-BOC protected
Vzorec: C₁₅H₃₂N₂O₂
MW: 272,427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02094501
CAS číslo: 216961-61-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR