3-[(Methylsulphonyl)amino]benzoic acid
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
3-(Methylsulphonamido)benzoic acid, N-(3-Carboxyphenyl)methanesulphonamide
OR10891-1G
OR10891-5G
APOSOR10891-1GEA
3060
CZK
APOSOR10891-1G
APOSOR10891-5G
3-[(Methylsulphonyl)amino]benzoic acid
3-[(Methylsulphonyl)amino]benzoic acid
Vzorec:
C₈H₉NO₄S MW: 215,226 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00596197 CAS číslo: 28547-13-9 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...