3-(Aminomethyl)-1H-indole oxalate
Dodavatel: Apollo Scientific
|
Varování
|
OR10823-500MG
OR10823-50MG
APOSOR10823-500MGEA
4140
CZK
APOSOR10823-500MG
APOSOR10823-50MG
3-(Aminomethyl)-1H-indole oxalate
3-(Aminomethyl)-1H-indole oxalate
Vzorec:
C₉H₁₀N₂·C₂H₂O₄ MW: 236.224 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01863232 CAS číslo: 296775-93-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...