Diethyl [N-(benzylidene)aminomethyl]phosphonate
Dodavatel: Apollo Scientific
|
Varování
|
OR10690-1G
OR10690-500MG
APOSOR10690-1GEA
3710
CZK
APOSOR10690-1G
APOSOR10690-500MG
Diethyl [N-(benzylidene)aminomethyl]phosphonate
Diethyl [N-(benzylidene)aminomethyl]phosphonate
Vzorec:
C₁₂H₁₈NO₃P MW: 255,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00152991 CAS číslo: 50917-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...