Tisk…

[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide 97%

Dodavatel: Apollo Scientific
Varování

OR10642-5G OR10642-1G OR10642-10G
APOSOR10642-5GEA 3110 CZK
APOSOR10642-5G APOSOR10642-1G APOSOR10642-10G
[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide 97%
[4-(Ethoxycarbonyl)butyl]triphenylphosphonium bromide
Vzorec: C₂₅H₂₈O₂P·Br
Storage Temperature: Ambient
MDL Number: MFCD00051866
CAS číslo: 118026-77-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR