Tisk…

(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid 96%

Dodavatel: Apollo Scientific

OR10357-1G OR10357-5G
APOSOR10357-1GEA 6350 CZK
APOSOR10357-1G APOSOR10357-5G
(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid 96%
(3-Benzyloxycarbonyl-5-nitrophenyl)boronic acid
Vzorec: C₁₄H₁₂BNO₆
Storage Temperature: Ambient
MDL Number: MFCD03411934
CAS číslo: 380430-62-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR