1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-[2-(Piperazin-1-yl)ethyl]-1H-pyrrole
OR0959-250MG
OR0959-1G
OR0959-5G
APOSOR0959-250MGEA
910
CZK
APOSOR0959-250MG
APOSOR0959-1G
APOSOR0959-5G
1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine
1-[2-(1H-Pyrrol-1-yl)ethyl]piperazine
Vzorec:
C₁₀H₁₇N₃ MW: 179,262 g/mol Bod varu: 96 °C (0,8 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD03412127 CAS číslo: 688763-20-4 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...