Tisk…

(Piperazin-1-yl)(pyrrolidin-1-yl)methanone 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-[(Piperazin-1-yl)carbonyl]pyrrolidine, 1-[(Pyrrolidin-1-yl)carbonyl]piperazine

OR0950-1G OR0950-5G OR0950-250MG
APOSOR0950-1GEA 2320 CZK
APOSOR0950-1G APOSOR0950-5G APOSOR0950-250MG
(Piperazin-1-yl)(pyrrolidin-1-yl)methanone 98%
(Piperazin-1-yl)(pyrrolidin-1-yl)methanone
Vzorec: C₉H₁₇N₃O
MW: 183,251 g/mol
Bod varu: 106 °C (0,015 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03844724
CAS číslo: 73331-93-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR