1-(3-Hydroxyprop-1-yl)-4-methylpiperazine 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(4-Methylpiperazin-1-yl)propan-1-ol
OR0881-1G
OR0881-5G
APOSOR0881-1GEA
0
CZK
APOSOR0881-1G
APOSOR0881-5G
1-(3-Hydroxyprop-1-yl)-4-methylpiperazine 95%
1-(3-Hydroxyprop-1-yl)-4-methylpiperazine
Vzorec:
C₈H₁₈N₂O Bod varu: 75…78 °C (0,5 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00009781 CAS číslo: 5317-33-9 EINECS: 226-177-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...