Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate
Dodavatel: Apollo Scientific
|
Varování
|
OR0880-10G
OR0880-1G
OR0880-5G
APOSOR0880-10GEA
52900
CZK
APOSOR0880-10G
APOSOR0880-1G
APOSOR0880-5G
Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate
Imidazo[2,1-b][1,3]benzothiazole-2-carboxylic acid hydrate
Vzorec:
C₁₀H₆N₂O₂S·H₂O Bod tání: 266…268 °C Storage Temperature: Ambient |
MDL Number:
MFCD08444056 CAS číslo: 1284226-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...