Tisk…

4,6-Dimethoxypyrimidine-2-carbonitrile 98%

Dodavatel: Apollo Scientific
Varování

OR0831-10G OR0831-5G OR0831-1G
APOSOR0831-10GEA 20500 CZK
APOSOR0831-10G APOSOR0831-5G APOSOR0831-1G
4,6-Dimethoxypyrimidine-2-carbonitrile 98%
4,6-Dimethoxypyrimidine-2-carbonitrile
Vzorec: C₇H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04972587
CAS číslo: 139539-63-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR