Tisk…

[3-(Morpholin-4-yl)phenyl]methylamine

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-(Morpholin-4-yl)benzylamine, 4-[3-(Aminomethyl)phenyl]morpholine

OR0717-1G OR0717-250MG
APOSOR0717-1GEA 8500 CZK
APOSOR0717-1G APOSOR0717-250MG
[3-(Morpholin-4-yl)phenyl]methylamine
[3-(Morpholin-4-yl)phenyl]methylamine
Vzorec: C₁₁H₁₆N₂O
MW: 192,258 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659079
CAS číslo: 625470-29-3
UN: 2735
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR