2-[(3,4-Dichlorophenoxy)methyl]oxirane
Dodavatel: Apollo Scientific
|
Varování
|
OR0431-1G
OR0431-250MG
APOSOR0431-1GEA
6950
CZK
APOSOR0431-1G
APOSOR0431-250MG
2-[(3,4-Dichlorophenoxy)methyl]oxirane
2-[(3,4-Dichlorophenoxy)methyl]oxirane
Vzorec:
C₉H₈Cl₂O₂ Bod varu: 170 °C (8 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00831502 CAS číslo: 21320-30-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...