Tisk…

3-(2,4-Dichlorophenyl)-1H-pyrazole

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 1,3-Dichloro-4-(1H-pyrazol-3-yl)benzene

OR0366-1G OR0366-250MG
APOSOR0366-1GEA 2480 CZK
APOSOR0366-1G APOSOR0366-250MG
3-(2,4-Dichlorophenyl)-1H-pyrazole
3-(2,4-Dichlorophenyl)-1H-pyrazole
Vzorec: C₉H₆Cl₂N₂
MW: 213,063 g/mol
Bod tání: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD01313364
CAS číslo: 154257-67-7
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR