(R)-Aminocarnitine
Dodavatel: Apollo Scientific
|
Varování
|
OR0340T-50MG
OR0340T-5MG
OR0340T-25MG
APOSOR0340T-50MGEA
55880
CZK
APOSOR0340T-50MG
APOSOR0340T-5MG
APOSOR0340T-25MG
(R)-Aminocarnitine
(R)-Aminocarnitine
Inhibits fatty acid oxidation. Acts as a hypoglycemic and antiketogenic compound and alterslipidic metabolism inhibiting carnitine acetyltransferase (CAT) and carnitinepalmitoyltransferase (CPT)
Vzorec:
C₇H₁₆N₂O₃ MW: 176,214 g/mol Bod tání: 153 °C Storage Temperature: Freezer |
MDL Number:
MFCD00797591 CAS číslo: 98063-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...