Benzo[b]furan-2-carbohydrazide
Dodavatel: Apollo Scientific
|
Varování
|
OR0275-1G
OR0275-250MG
APOSOR0275-1GEA
2780
CZK
APOSOR0275-1G
APOSOR0275-250MG
Benzo[b]furan-2-carbohydrazide
Benzo[b]furan-2-carbohydrazide
Vzorec:
C₉H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00456546 CAS číslo: 42974-19-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...