Tisk…

1-(4-Nitrobenzyl)piperazine dihydrochloride 97%

Dodavatel: Apollo Scientific

Synonyms: 4-[(Piperazin-1-yl)methyl]nitrobenzene dihydrochloride

OR0227-10G OR0227-2G OR0227-5G
APOSOR0227-10GEA 12170 CZK
APOSOR0227-10G APOSOR0227-2G APOSOR0227-5G
1-(4-Nitrobenzyl)piperazine dihydrochloride 97%
1-(4-Nitrobenzyl)piperazine dihydrochloride
Vzorec: C₁₁H₁₅N₃O₂·2ClH
MW: 294,178 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04972581
CAS číslo: 422517-67-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR