Tisk…

1-(2,4-Dichlorophenyl)biguanide hydrochloride

Dodavatel: Apollo Scientific
Varování

Synonyms: N-(2,4-Dichlorophenyl)imidodicarbonimidic diamide hydrochloride

OR0193-100G OR0193-25G OR0193-5G
APOSOR0193-100GEA 10650 CZK
APOSOR0193-100G APOSOR0193-25G APOSOR0193-5G
1-(2,4-Dichlorophenyl)biguanide hydrochloride
1-(2,4-Dichlorophenyl)biguanide hydrochloride
Vzorec: C₈H₉Cl₂N₅·ClH
MW: 282,558 g/mol
Bod tání: 259,5…263.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00203356
CAS číslo: 6634-65-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR