Tisk…

6-Chloro-2-(piperazin-1-yl)-1,3-benzothiazole 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(6-Chloro-1,3-benzothiazol-2-yl)piperazine

OR018290-1G OR018290-250MG
APOSOR018290-1GEA 8270 CZK
APOSOR018290-1G APOSOR018290-250MG
6-Chloro-2-(piperazin-1-yl)-1,3-benzothiazole 95+%
6-Chloro-2-(piperazin-1-yl)-1,3-benzothiazole
Vzorec: C₁₁H₁₂ClN₃S
MW: 253,751 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01571353
CAS číslo: 153025-29-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR