Tisk…

2-Phenyl-1H-benzimidazole-7-carboxylic acid 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 7-Carboxy-2-phenyl-1H-benzimidazole, 2-Phenyl-3H-benzimidazole-4-carboxylic acid

OR01760-500MG OR01760-250MG
APOSOR01760-500MGEA 7670 CZK
APOSOR01760-500MG APOSOR01760-250MG
2-Phenyl-1H-benzimidazole-7-carboxylic acid 95%
2-Phenyl-1H-benzimidazole-7-carboxylic acid
Vzorec: C₁₄H₁₀N₂O₂
MW: 238,241 g/mol
Bod tání: 297…300 °C
Storage Temperature: Ambient
MDL Number: MFCD06245480
CAS číslo: 66630-72-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR