Tisk…

3-Butoxycyclohex-2-en-1-one

Dodavatel: Apollo Scientific

OR0176-10G OR0176-1G OR0176-5G
APOSOR0176-10GEA 11250 CZK
APOSOR0176-10G APOSOR0176-1G APOSOR0176-5G
3-Butoxycyclohex-2-en-1-one
3-Butoxycyclohex-2-en-1-one
Vzorec: C₁₀H₁₆O₂
Bod varu: 254 °C
Storage Temperature: Ambient
MDL Number: MFCD00190682
CAS číslo: 16493-04-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR