Tisk…

2-Phenyl-3H-benzimidazole-5-carboxylic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Phenyl-1H-benzimidazole-6-carboxylic acid

OR01759-100MG OR01759-1G OR01759-250MG
APOSOR01759-100MGEA 1660 CZK
APOSOR01759-100MG APOSOR01759-1G APOSOR01759-250MG
2-Phenyl-3H-benzimidazole-5-carboxylic acid
2-Phenyl-3H-benzimidazole-5-carboxylic acid
Vzorec: C₁₄H₁₀N₂O₂
MW: 238,241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245479
CAS číslo: 66630-70-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR