Tisk…

6-(3-Phenylpiperazin-1-yl)nicotinic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Carboxy-2-(3-phenylpiperazin-1-yl)pyridine, 6-(3-Phenylpiperazin-1-yl)pyridine-3-carboxylic acid

OR01726-100MG OR01726-50MG
APOSOR01726-100MGEA 4140 CZK
APOSOR01726-100MG APOSOR01726-50MG
6-(3-Phenylpiperazin-1-yl)nicotinic acid
6-(3-Phenylpiperazin-1-yl)nicotinic acid
Vzorec: C₁₆H₁₇N₃O₂
MW: 283,325 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06245561
CAS číslo: 904816-46-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR