Tisk…

5-(tert-Butyl)thiophene-2-carboxylic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 5-(tert-Butyl)-2-thenoic acid, 5-(tert-Butyl)-2-carboxythiophene

OR01670-1G OR01670-5G
APOSOR01670-1GEA 1290 CZK
APOSOR01670-1G APOSOR01670-5G
5-(tert-Butyl)thiophene-2-carboxylic acid
5-(tert-Butyl)thiophene-2-carboxylic acid
Vzorec: C₉H₁₂O₂S
MW: 184,255 g/mol
Bod tání: 125…127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01566583
CAS číslo: 29212-25-7
UN: 1759
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR