4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR015022-250MG
OR015022-100MG
OR015022-1G
APOSOR015022-250MGEA
3540
CZK
APOSOR015022-250MG
APOSOR015022-100MG
APOSOR015022-1G
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(oxetan-3-yl)-1,3,2-dioxaborolane
Vzorec:
C₉H₁₇BO₃ MW: 184,04 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD26521383 CAS číslo: 1396215-84-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...