Tisk…

2-Acetyl-3H-benzo[f]chromen-3-one

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Acetyl-3-oxo-3H-benzo[f]chromene

OR0089-1G OR0089-500MG OR0089-5G
APOSOR0089-1GEA 2480 CZK
APOSOR0089-1G APOSOR0089-500MG APOSOR0089-5G
2-Acetyl-3H-benzo[f]chromen-3-one
2-Acetyl-3H-benzo[f]chromen-3-one
Vzorec: C₁₅H₁₀O₃
MW: 238,238 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00445975
CAS číslo: 727-80-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR