1-(Biphenyl-2-yl)piperazine dihydrochloride 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Piperazin-1-yl)biphenyl dihydrochloride
OR0078-1G
OR0078-500MG
APOSOR0078-1GEA
4630
CZK
APOSOR0078-1G
APOSOR0078-500MG
1-(Biphenyl-2-yl)piperazine dihydrochloride 97%
1-(Biphenyl-2-yl)piperazine dihydrochloride
Vzorec:
C₁₆H₁₈N₂·2ClH MW: 311.249 g/mol Bod tání: 205…209 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094588 CAS číslo: 769944-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...