Tisk…

Niobium(I) hydride 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Niobium hydride

IN2755-25G IN2755-5G
APOSIN2755-25GEA 0 CZK
APOSIN2755-25G APOSIN2755-5G
Niobium(I) hydride 99%
Niobium(I) hydride
Vzorec: H₅Nb
MW: 97.946 g/mol
Bod tání: 500 °C (rozkl.)
Hustota: 6.6 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS číslo: 13981-86-7
UN: 3132
ADR: 4.3+4.1 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR