Tisk…
Tenuazonic acid, copper salt 99%
Dodavatel: Apollo Scientific
Varování
BIT1600-10MG BIT1600-1MG BIT1600-5MG
APOSBIT1600-10MGEA 15640 CZK
APOSBIT1600-10MG APOSBIT1600-1MG APOSBIT1600-5MG
Tenuazonic acid, copper salt 99%
Tenuazonic acid, copper salt
Vzorec: Cu(C₁₀H₁₄NO₃)₂
Storage Temperature: Refrigerator
MDL Number: MFCD00083505
UN: 2811
ADR: 6.1,III