Thiol-PEG3-acid
Dodavatel: Apollo Scientific
BIPG1837-250MG
BIPG1837-1G
BIPG1837-500MG
APOSBIPG1837-250MGEA
6850
CZK
APOSBIPG1837-250MG
APOSBIPG1837-1G
APOSBIPG1837-500MG
Thiol-PEG3-acid
Thiol-PEG3-acid
Vzorec:
C₉H₁₈O₅S Storage Temperature: Ambient |
MDL Number:
MFCD25424073 CAS číslo: 1347750-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...