Tisk…

Thiol-PEG2-acid

Dodavatel: Apollo Scientific

BIPG1836-250MG BIPG1836-500MG
APOSBIPG1836-250MGEA 10350 CZK
APOSBIPG1836-250MG APOSBIPG1836-500MG
Thiol-PEG2-acid
Thiol-PEG2-acid
Vzorec: C₇H₁₄O₄S
MW: 194,249 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24503001
CAS číslo: 1379649-73-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR