Tisk…

t-Boc-N-Amido-PEG2-Azide 98%

Dodavatel: Apollo Scientific

BIPG1820-1G BIPG1820-5G BIPG1820-250MG
APOSBIPG1820-1GEA 5590 CZK
APOSBIPG1820-1G APOSBIPG1820-5G APOSBIPG1820-250MG
t-Boc-N-Amido-PEG2-Azide 98%
t-Boc-N-Amido-PEG2-Azide
Vzorec: C₁₁H₂₂N₄O₄
MW: 274,317 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23380094
CAS číslo: 950683-55-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR