Tisk…

t-Boc-N-Amido-PEG12-acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Boc-NH-PEG12-CH2CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11,14,17,20,23,26,29,32,35,38,41-tridecaoxa-5-azatetratetracontan-44-oic acid

BIPG1807-250MG BIPG1807-50MG BIPG1807-100MG
APOSBIPG1807-250MGEA 15510 CZK
APOSBIPG1807-250MG APOSBIPG1807-50MG APOSBIPG1807-100MG
t-Boc-N-Amido-PEG12-acid
t-Boc-N-Amido-PEG12-acid
Vzorec: C₃₂H₆₃NO₁₆
Storage Temperature: Refrigerator
MDL Number: MFCD13185149
CAS číslo: 1415981-79-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR