Tisk…

t-Boc-N-Amido-PEG5-acid

Dodavatel: Apollo Scientific
Varování

Synonyms: Boc-NH-PEG5-CH2CH2COOH, 2,2-Dimethyl-4-oxo-3,8,11,14,17,20-hexaoxa-5-azatricosan-23-oic acid

BIPG1803-100MG BIPG1803-500MG BIPG1803-1G
APOSBIPG1803-100MGEA 2690 CZK
APOSBIPG1803-100MG APOSBIPG1803-500MG APOSBIPG1803-1G
t-Boc-N-Amido-PEG5-acid
t-Boc-N-Amido-PEG5-acid
Vzorec: C₁₈H₃₅NO₉
MW: 409,472 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24539478
CAS číslo: 1347750-78-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR