Tisk…

Propargyl-PEG3-Bromide

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(2-(2-(2-Bromoethoxy)ethoxy)ethoxy)prop-1-yne

BIPG1729-250MG BIPG1729-100MG
APOSBIPG1729-250MGEA 18620 CZK
APOSBIPG1729-250MG APOSBIPG1729-100MG
Propargyl-PEG3-Bromide
Propargyl-PEG3-Bromide
Vzorec: C₉H₁₅BrO₃
Storage Temperature: Freezer
MDL Number: MFCD28505517
CAS číslo: 203740-63-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR