Mal-PEG8-acid
Dodavatel: Apollo Scientific
BIPG1497-100MG
BIPG1497-50MG
APOSBIPG1497-100MGEA
19080
CZK
APOSBIPG1497-100MG
APOSBIPG1497-50MG
Mal-PEG8-acid
Mal-PEG8-acid
Vzorec:
C₂₃H₃₉NO₁₂ MW: 521,555 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD26793771 CAS číslo: 1818294-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...