Tisk…

Hydroxy-PEG1-CH2CO2tBu

Dodavatel: Apollo Scientific
Varování

Synonyms: tert-Butyl 2-(2-hydroxyethoxy)acetate, 2-Methyl-2-propanyl (2-hydroxyethoxy)acetate, Hydroxy-PEG1-CH2-Boc

BIPG1435-250MG
APOSBIPG1435-250MGEA 622 CZK
APOSBIPG1435-250MG
Hydroxy-PEG1-CH2CO2tBu
Hydroxy-PEG1-CH2CO2tBu
Vzorec: C₈H₁₆O₄
MW: 176,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27635155
CAS číslo: 287174-32-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR